Assay Details
| Gene Symbol: KCNH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001204798.1 | NP_001191727.1 | 1 | 271 | 95 | |
| NM_172057.2 | NP_742054.1 | 1 | 271 | 95 | ||
| GenBank mRNA | BC127673.1 | - | 1 | 260 | 95 | |
| BG683210.1 | - | 1 | 271 | 95 | ||
| BQ890248.1 | - | 1 | 269 | 95 | ||
| DQ120125.1 | - | 1 | 348 | 95 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC127673.1, DQ120125.1, NM_001204798.1, NM_172057.2 transcripts.
Set Membership: |
|

KCNH2
Genomic Map