Assay Details
| Gene Symbol: SLC20A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006716391.3 | XP_006716454.1 | 1 | 739 | 91 | |
| XM_017013749.1 | XP_016869238.1 | 1 | 739 | 91 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006716391.3, XM_017013749.1 transcripts.
Set Membership: |
|

SLC20A2
Genomic Map