Assay Details
| Gene Symbol: FLJ37453 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_024279.1 | 2 | 138 | 92 | ||
| GenBank mRNA | AK124018.1 | - | 2 | 138 | 92 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

FLJ37453| Gene Symbol: FLJ37453 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_024279.1 | 2 | 138 | 92 | ||
| GenBank mRNA | AK124018.1 | - | 2 | 138 | 92 | |
Set Membership: |
|