Assay Details
| Gene Symbol: SEC62-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AV661681.1 | - | 1 | 217 | 115 | |
| AV661720.1 | - | 1 | 217 | 115 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SEC62-AS1| Gene Symbol: SEC62-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AV661681.1 | - | 1 | 217 | 115 | |
| AV661720.1 | - | 1 | 217 | 115 | ||
Set Membership: |
|