Assay Details
| Gene Symbol: SNORD53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_002741.2 | 1 | 38 | 68 | ||
| GenBank mRNA | X96652.1 | - | 1 | 38 | 68 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
small nucleolar RNA, C/D box 92 |
SNORD92 | HBII-316 | -13.3kb | Sense | Chr. 2 - 28913662 - 28913750 on NCBI Build GRCh38 |
| Downstream | family with sequence similarity 179 member A | FAM179A | 54.2kb | Sense | Chr. 2 - 28981298 - 29052230 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

SNORD53
Genomic Map