Assay Details
| Gene Symbol: SPRR2G | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014291.3 | NP_001014313.1 | 2 | 103 | ||
| XM_017002177.1 | XP_016857666.1 | 4 | 103 | |||
| XM_017002178.1 | XP_016857667.1 | 2 | 103 | |||
| GenBank mRNA | AA456642.1 | - | 1 | 434 | 103 | |
| AA490630.1 | - | 1 | 103 | |||
| BC130455.1 | - | 2 | 103 | |||
| BC133041.1 | - | 2 | 103 | |||
More Information
Set Membership: |
|

SPRR2G
Genomic Map