Assay Details
| Gene Symbol: SCN7A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_002976.3 | NP_002967.2 | 25 | 5089 | 84 | |
| NR_045628.1 | 26 | 5183 | 84 | |||
| XM_006712680.2 | XP_006712743.1 | 25 | 5158 | 84 | ||
| XM_006712681.3 | XP_006712744.1 | 26 | 5383 | 84 | ||
| XM_006712682.3 | XP_006712745.1 | 27 | 5508 | 84 | ||
| XM_011511615.2 | XP_011509917.1 | 27 | 5454 | 84 | ||
| XM_017004667.1 | XP_016860156.1 | 27 | 5427 | 84 | ||
| GenBank mRNA | BC141864.1 | - | 26 | 5185 | 84 | |
| M91556.1 | - | 25 | 5098 | 84 | ||
| S75992.1 | - | 1 | 122 | 84 | ||
More Information
Set Membership: |
|

SCN7A
Genomic Map