Assay Details
| Gene Symbol: PPARD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001171818.1 | NP_001165289.1 | 9 | 2658 | 99 | |
| NM_001171819.1 | NP_001165290.1 | 7 | 2331 | 99 | ||
| NM_001171820.1 | NP_001165291.1 | 6 | 2268 | 99 | ||
| NM_006238.4 | NP_006229.1 | 8 | 2562 | 99 | ||
| XM_005249193.1 | XP_005249250.1 | 8 | 2577 | 99 | ||
| XM_006715120.1 | XP_006715183.1 | 10 | 2715 | 99 | ||
| XM_006715123.1 | XP_006715186.1 | 7 | 2465 | 99 | ||
| XM_011514707.1 | XP_011513009.1 | 10 | 3312 | 99 | ||
| XM_011514709.1 | XP_011513011.1 | 10 | 3286 | 99 | ||
| XM_011514710.1 | XP_011513012.1 | 9 | 2639 | 99 | ||
| XM_017010971.1 | XP_016866460.1 | 11 | 3408 | 99 | ||
| XM_017010972.1 | XP_016866461.1 | 10 | 3339 | 99 | ||
| XM_017010973.1 | XP_016866462.1 | 10 | 2774 | 99 | ||
| XM_017010974.1 | XP_016866463.1 | 9 | 2699 | 99 | ||
| XM_017010975.1 | XP_016866464.1 | 7 | 2334 | 99 | ||
| GenBank mRNA | AK122614.1 | - | 8 | 2555 | 99 | |
| BC127809.1 | - | 1 | 123 | 99 | ||
More Information
Set Membership: |
|

PPARD
Genomic Map