Assay Details
| Gene Symbol: MRPL12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_002949.3 | NP_002940.2 | 2 | 292 | 93 | |
| GenBank mRNA | BC002344.2 | - | 2 | 277 | 2961288 | 93 |
| BC007497.2 | - | 2 | 174 | 2959986 | 93 | |
| DA648796.1 | - | 2 | 292 | 93 | ||
| EF036488.1 | - | 2 | 162 | 93 | ||
| U25041.1 | - | 2 | 178 | 93 | ||
| X79865.1 | - | 2 | 297 | 93 | ||
More Information
Set Membership: |
|

MRPL12
Genomic Map