Assay Details
| Gene Symbol: P3H4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_006455.2 | NP_006446.1 | 1 | 648 | 94 | |
| XM_006721640.3 | XP_006721703.1 | 1 | 1066 | 94 | ||
| GenBank mRNA | AJ250583.1 | - | 1 | 449 | 94 | |
| AK056085.1 | - | 2 | 831 | 94 | ||
| AK222945.1 | - | 1 | 393 | 94 | ||
| AY295765.1 | - | 1 | 116 | 94 | ||
| BC001047.2 | - | 1 | 401 | 3510406 | 94 | |
| BC007942.2 | - | 1 | 396 | 4130726 | 94 | |
| BC011701.2 | - | 1 | 380 | 3510378 | 94 | |
| U47621.1 | - | 1 | 375 | 94 | ||
More Information
Set Membership: |
|

P3H4
Genomic Map