Assay Details
| Gene Symbol: DDX21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_004728.3 | NP_004719.2 | 1 | 69 | 97 | |
| XM_011540336.1 | XP_011538638.1 | 1 | 61 | 97 | ||
| GenBank mRNA | BC104671.1 | - | 1 | 68 | 97 | |
| BU963561.1 | - | 1 | 67 | 97 | ||
| DW408689.1 | - | 1 | 86 | 97 | ||
| Gene Symbol: DDX50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017016626.1 | XP_016872115.1 | 16 | 4285 | 97 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC104671.1, NM_004728.3, XM_011540336.1 transcripts.
Set Membership: |
|

DDX21
Genomic Map