Assay Details
| Gene Symbol: CLPP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_006012.2 | NP_006003.1 | 1 | 162 | 91 | |
| GenBank mRNA | AK311973.1 | - | 1 | 62 | 91 | |
| BC002956.1 | - | 1 | 162 | 3542292 | 91 | |
| EF036491.1 | - | 1 | 56 | 91 | ||
| Z50853.1 | - | 1 | 58 | 91 | ||
More Information
Set Membership: |
|

CLPP
Genomic Map