Assay Details
| Gene Symbol: MED25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF447873.1 | - | 4 | 2855 | 69 | |
| AJ617479.1 | - | 18 | 2185 | 69 | ||
| AK055844.1 | - | 18 | 2226 | 69 | ||
| AL136746.1 | - | 18 | 2186 | 69 | ||
| EU392499.1 | - | 2 | 350 | 69 | ||
| EU392515.1 | - | 4 | 389 | 69 | ||
More Information
Set Membership: |
|

MED25
Genomic Map