Assay Details
| Gene Symbol: MED16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_005481.2 | NP_005472.2 | 9 | 1626 | 82 | |
| XM_017026121.1 | XP_016881610.1 | 8 | 1466 | 82 | ||
| GenBank mRNA | AF106934.1 | - | 9 | 1613 | 82 | |
| AF121228.1 | - | 8 | 1475 | 82 | ||
| AL137732.1 | - | 2 | 328 | 82 | ||
| BC004554.1 | - | 9 | 1607 | 4053743 | 82 | |
| BC007853.2 | - | 5 | 736 | 82 | ||
| BC011841.1 | - | 9 | 1607 | 4053664 | 82 | |
| BC017282.2 | - | 9 | 1601 | 4876977 | 82 | |
| EU392451.1 | - | 7 | 1054 | 82 | ||
| EU392452.1 | - | 5 | 775 | 82 | ||
| EU392453.1 | - | 5 | 769 | 82 | ||
| EU392454.1 | - | 5 | 646 | 82 | ||
| EU392455.1 | - | 3 | 211 | 82 | ||
| EU392485.1 | - | 4 | 240 | 82 | ||
More Information
Set Membership: |
|

MED16
Genomic Map