Assay Details
| Gene Symbol: PTPN22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193431.2 | NP_001180360.1 | 8 | 722 | 86 | |
| NM_001308297.1 | NP_001295226.1 | 7 | 650 | 86 | ||
| NM_012411.5 | NP_036543.4 | 8 | 722 | 86 | ||
| NM_015967.6 | NP_057051.3 | 8 | 722 | 86 | ||
| XM_011541221.1 | XP_011539523.1 | 8 | 703 | 86 | ||
| XM_011541222.1 | XP_011539524.1 | 8 | 703 | 86 | ||
| XM_011541223.2 | XP_011539525.1 | 8 | 703 | 86 | ||
| XM_011541225.2 | XP_011539527.1 | 7 | 652 | 86 | ||
| XM_017001004.1 | XP_016856493.1 | 8 | 703 | 86 | ||
| XM_017001005.1 | XP_016856494.1 | 8 | 684 | 86 | ||
| XM_017001006.1 | XP_016856495.1 | 8 | 703 | 86 | ||
| GenBank mRNA | AF001846.1 | - | 8 | 627 | 86 | |
| AF077031.1 | - | 8 | 658 | 86 | ||
| AF150732.1 | - | 8 | 656 | 86 | ||
| AK310698.1 | - | 8 | 675 | 86 | ||
| BC071670.1 | - | 8 | 661 | 5497108 | 86 | |
| GU479452.1 | - | 8 | 636 | 86 | ||
| JN084012.1 | - | 7 | 527 | 86 | ||
| U69700.1 | - | 2 | 110 | 86 | ||
More Information
Set Membership: |
|

PTPN22
Genomic Map