Assay Details
| Gene Symbol: REN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000537.3 | NP_000528.1 | 2 | 255 | 67 | |
| GenBank mRNA | AF117822.1 | - | 2 | 184 | 67 | |
| BC033474.1 | - | 2 | 237 | 5188566 | 67 | |
| BC047752.1 | - | 2 | 240 | 5590211 | 67 | |
| BP321283.1 | - | 2 | 255 | 67 | ||
| CR536498.1 | - | 2 | 67 | |||
More Information
Set Membership: |
|

REN
Genomic Map