Assay Details
| Gene Symbol: NEK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001204182.1 | NP_001191111.1 | 3 | 631 | 102 | |
| NM_001204183.1 | NP_001191112.1 | 3 | 631 | 102 | ||
| NM_002497.3 | NP_002488.1 | 3 | 631 | 102 | ||
| XM_005273147.1 | XP_005273204.1 | 3 | 628 | 102 | ||
| GenBank mRNA | AK223353.1 | - | 3 | 592 | 102 | |
| AK225522.1 | - | 3 | 592 | 102 | ||
| AK303092.1 | - | 4 | 494 | 102 | ||
| AY045701.1 | - | 3 | 562 | 102 | ||
| BC043502.1 | - | 3 | 619 | 5541026 | 102 | |
| BC052807.1 | - | 3 | 626 | 102 | ||
| BC065932.1 | - | 3 | 621 | 102 | ||
| BT019729.1 | - | 3 | 480 | 102 | ||
| U11050.1 | - | 3 | 614 | 102 | ||
| Z25425.1 | - | 2 | 234 | 102 | ||
| Z29066.1 | - | 3 | 562 | 102 | ||
More Information
Set Membership: |
|

NEK2
Genomic Map