Assay Details
| Gene Symbol: B3GALT5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017028227.1 | XP_016883716.1 | 1 | 1426 | 60 | |
| XM_017028228.1 | XP_016883717.1 | 1 | 1426 | 60 | ||
| XM_017028229.1 | XP_016883718.1 | 1 | 1426 | 60 | ||
| XM_017028230.1 | XP_016883719.1 | 1 | 1426 | 60 | ||
| XM_017028231.1 | XP_016883720.1 | 1 | 1426 | 60 | ||
| XM_017028232.1 | XP_016883721.1 | 1 | 1426 | 60 | ||
| XM_017028238.1 | XP_016883727.1 | 1 | 1426 | 60 | ||
| XM_017028239.1 | XP_016883728.1 | 1 | 1426 | 60 | ||
| XM_017028240.1 | XP_016883729.1 | 1 | 1426 | 60 | ||
| GenBank mRNA | DQ645737.1 | - | 1 | 44 | 60 | |
| DQ645738.1 | - | 1 | 44 | 60 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017028227.1, XM_017028228.1, XM_017028229.1, XM_017028230.1, XM_017028231.1, XM_017028232.1, XM_017028238.1, XM_017028239.1, XM_017028240.1 transcripts.
Set Membership: |
|

B3GALT5
Genomic Map