Assay Details
| Gene Symbol: MX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144925.2 | NP_001138397.1 | 18 | 2534 | 89 | |
| NM_001178046.2 | NP_001171517.1 | 14 | 1918 | 89 | ||
| NM_002462.4 | NP_002453.2 | 16 | 2093 | 89 | ||
| XM_005260978.4 | XP_005261035.1 | 16 | 2009 | 89 | ||
| XM_005260979.2 | XP_005261036.1 | 16 | 2202 | 89 | ||
| XM_005260980.2 | XP_005261037.1 | 16 | 1974 | 89 | ||
| XM_005260981.2 | XP_005261038.1 | 15 | 1854 | 89 | ||
| XM_005260982.2 | XP_005261039.1 | 15 | 1839 | 89 | ||
| XM_011529568.2 | XP_011527870.1 | 20 | 2543 | 89 | ||
| XM_017028349.1 | XP_016883838.1 | 18 | 2553 | 89 | ||
| XM_017028350.1 | XP_016883839.1 | 16 | 2221 | 89 | ||
| XM_017028351.1 | XP_016883840.1 | 15 | 1877 | 89 | ||
| XM_017028352.1 | XP_016883841.1 | 15 | 1850 | 89 | ||
| GenBank mRNA | AK093008.1 | - | 12 | 1841 | 89 | |
| AK096355.1 | - | 18 | 2577 | 89 | ||
| AK315465.1 | - | 14 | 1775 | 89 | ||
| AY186254.1 | - | 2 | 97 | 89 | ||
| BC014222.2 | - | 11 | 1302 | 89 | ||
| BC032602.1 | - | 15 | 1843 | 5535278 | 89 | |
| M30817.1 | - | 14 | 1762 | 89 | ||
| M33882.1 | - | 16 | 1897 | 89 | ||
More Information
Set Membership: |
|

MX1
Genomic Map