Assay Details
| Gene Symbol: DNAH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015512.4 | NP_056327.4 | 29 | 5027 | 98 | |
| XM_017006129.1 | XP_016861618.1 | 30 | 7915 | 98 | ||
| XM_017006130.1 | XP_016861619.1 | 30 | 7914 | 98 | ||
| XM_017006131.1 | XP_016861620.1 | 30 | 7915 | 98 | ||
| XM_017006132.1 | XP_016861621.1 | 30 | 7916 | 98 | ||
| XM_017006133.1 | XP_016861622.1 | 30 | 7916 | 98 | ||
| GenBank mRNA | AB037831.3 | - | 29 | 5027 | 98 | |
| AB290163.1 | - | 29 | 5027 | 98 | ||
| AJ132083.1 | - | 2 | 140 | 98 | ||
More Information
Set Membership: |
|

DNAH1
Genomic Map