Assay Details
| Gene Symbol: CXCL6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_002993.3 | NP_002984.1 | 2 | 363 | 69 | |
| GenBank mRNA | AK311981.1 | - | 2 | 240 | 69 | |
| BC013744.1 | - | 2 | 198 | 4471095 | 69 | |
| BP204144.1 | - | 2 | 363 | 69 | ||
| U81234.1 | - | 2 | 231 | 69 | ||
| Y08770.1 | - | 1 | 57 | 69 | ||
More Information
Set Membership: |
|

CXCL6
Genomic Map