Assay Details
| Gene Symbol: HNRNPAB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_004499.3 | NP_004490.2 | 6 | 979 | 67 | |
| NM_031266.2 | NP_112556.2 | 6 | 979 | 67 | ||
| GenBank mRNA | AK054600.1 | - | 5 | 1027 | 67 | |
| AK097657.1 | - | 6 | 980 | 67 | ||
| AK223425.1 | - | 6 | 956 | 67 | ||
| BC001616.1 | - | 6 | 938 | 3547193 | 67 | |
| BC002625.2 | - | 6 | 938 | 3606731 | 67 | |
| BC004561.2 | - | 6 | 946 | 3960088 | 67 | |
| BC009359.2 | - | 6 | 942 | 4301401 | 67 | |
| BC036708.2 | - | 6 | 905 | 4331453 | 67 | |
| M65028.1 | - | 6 | 861 | 67 | ||
| U76713.1 | - | 5 | 734 | 67 | ||
| Z36844.1 | - | 2 | 147 | 67 | ||
More Information
Set Membership: |
|

HNRNPAB
Genomic Map