Assay Details
| Gene Symbol: TH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199292.2 | NP_954986.2 | 2 | 165 | 61 | |
| NM_199293.2 | NP_954987.2 | 2 | 153 | 61 | ||
| GenBank mRNA | M17589.1 | - | 2 | 155 | 61 | |
| M24790.1 | - | 2 | 165 | 61 | ||
| Y00414.1 | - | 2 | 147 | 61 | ||
More Information
Set Membership: |
|

TH
Genomic Map