Assay Details
| Gene Symbol: MMP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_002422.4 | NP_002413.1 | 1 | 217 | 64 | |
| GenBank mRNA | AK223283.1 | - | 1 | 135 | 64 | |
| AK223291.1 | - | 1 | 136 | 64 | ||
| AK313310.1 | - | 1 | 133 | 64 | ||
| BC069676.1 | - | 1 | 137 | 7262578 | 64 | |
| BC069716.1 | - | 1 | 137 | 7262590 | 64 | |
| BC074815.2 | - | 1 | 119 | 30915319 | 64 | |
| BC074869.2 | - | 1 | 119 | 30915468 | 64 | |
| BC105954.1 | - | 1 | 130 | 40033171 | 64 | |
| BC107490.1 | - | 1 | 130 | 40033168 | 64 | |
| BC107491.1 | - | 1 | 130 | 40033170 | 64 | |
| DA982602.1 | - | 1 | 217 | 64 | ||
| DQ399598.1 | - | 1 | 38 | 64 | ||
| J03209.1 | - | 1 | 74 | 64 | ||
| X05232.1 | - | 1 | 117 | 64 | ||
More Information
Set Membership: |
|

MMP3
Genomic Map