Assay Details
| Gene Symbol: TP53AIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195195.1 | NP_001182124.1 | 1 | 564 | 60 | |
| NM_001251964.1 | NP_001238893.1 | 1 | 564 | 60 | ||
| NM_022112.2 | NP_071395.2 | 1 | 564 | 60 | ||
| XM_017018113.1 | XP_016873602.1 | 1 | 1389 | 60 | ||
| XM_017018114.1 | XP_016873603.1 | 1 | 1395 | 60 | ||
| XM_017018115.1 | XP_016873604.1 | 1 | 1398 | 60 | ||
| XM_017018117.1 | XP_016873606.1 | 1 | 1394 | 60 | ||
| GenBank mRNA | AB045830.1 | - | 1 | 92 | 60 | |
| AB045831.1 | - | 1 | 92 | 60 | ||
| AB045832.1 | - | 1 | 92 | 60 | ||
| AK310612.1 | - | 1 | 310 | 60 | ||
| AK314286.1 | - | 1 | 308 | 60 | ||
| AL703609.1 | - | 1 | 564 | 60 | ||
| DB111427.1 | - | 1 | 308 | 60 | ||
| DB129822.1 | - | 1 | 310 | 60 | ||
| EF693749.1 | - | 1 | 68 | 60 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AB045830.1, AB045831.1, AB045832.1, AK314286.1, EF693749.1, NM_001195195.1, NM_001251964.1, NM_022112.2, XM_017018113.1, XM_017018114.1, XM_017018115.1, XM_017018117.1 transcripts.
Set Membership: |
|

TP53AIP1
Genomic Map