Assay Details
| Gene Symbol: GCNT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145655.3 | NP_663630.2 | 1 | 668 | 96 | |
| XM_011514466.2 | XP_011512768.1 | 1 | 1112 | 96 | ||
| GenBank mRNA | AB078433.1 | - | 1 | 438 | 96 | |
| AF458026.1 | - | 1 | 438 | 96 | ||
| AF458027.1 | - | 1 | 438 | 96 | ||
| AF458028.1 | - | 1 | 438 | 96 | ||
| AF458029.1 | - | 1 | 438 | 96 | ||
| AF458030.1 | - | 1 | 438 | 96 | ||
| AK313426.1 | - | 1 | 683 | 96 | ||
| AL832719.1 | - | 1 | 669 | 96 | ||
| AY435147.1 | - | 1 | 438 | 96 | ||
| BC130524.1 | - | 1 | 610 | 40146555 | 96 | |
| BX503586.1 | - | 1 | 95 | |||
| BX647576.1 | - | 1 | 667 | 96 | ||
| BX647668.1 | - | 1 | 669 | 96 | ||
More Information
Set Membership: |
|

GCNT2
Genomic Map