Assay Details
| Gene Symbol: MED23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270522.1 | NP_001257451.1 | 30 | 4375 | 148 | |
| NM_004830.3 | NP_004821.2 | 29 | 5146 | 148 | ||
| XM_005267223.2 | XP_005267280.1 | 30 | 5160 | 148 | ||
| XM_011536257.2 | XP_011534559.1 | 28 | 5046 | 148 | ||
| XM_017011501.1 | XP_016866990.1 | 20 | 4104 | 148 | ||
| GenBank mRNA | AB033042.1 | - | 29 | 5115 | 148 | |
| AF105332.1 | - | 29 | 5076 | 148 | ||
| AI249380.1 | - | 1 | 148 | |||
| AJ420587.1 | - | 7 | 3092 | 148 | ||
| EU392521.1 | - | 6 | 791 | 148 | ||
| EU392524.1 | - | 3 | 423 | 148 | ||
| EU392525.1 | - | 2 | 983 | 148 | ||
More Information
Set Membership: |
|

MED23
Genomic Map