Assay Details
| Gene Symbol: SOX17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022454.3 | NP_071899.1 | 1 | 332 | 110 | |
| GenBank mRNA | AB073988.1 | - | 1 | 131 | 110 | |
| AK025905.1 | - | 1 | 332 | 110 | ||
| BC030209.1 | - | 1 | 320 | 110 | ||
| BC111365.1 | - | 1 | 288 | 110 | ||
| DA874945.1 | - | 1 | 330 | 110 | ||
More Information
Set Membership: |
|

SOX17
Genomic Map