Assay Details
| Gene Symbol: RTL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001134888.2 | NP_001128360.1 | 1 | 2872 | 81 | |
| GenBank mRNA | BC150617.1 | - | 1 | 2872 | 9056987 | 81 |
More Information
Set Membership: |
|

RTL1
Genomic Map