Assay Details
| Gene Symbol: BCL2A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114735.1 | NP_001108207.1 | 1 | 479 | 152 | |
| NM_004049.3 | NP_004040.1 | 1 | 479 | 152 | ||
| GenBank mRNA | AL110097.1 | - | 1 | 347 | 152 | |
| AY234180.1 | - | 1 | 305 | 152 | ||
| BC016281.1 | - | 1 | 357 | 3920808 | 152 | |
| BG198875.1 | - | 1 | 625 | 153 | ||
| BG216703.1 | - | 1 | 480 | 152 | ||
| BT007103.1 | - | 1 | 297 | 152 | ||
| CD640106.1 | - | 1 | 245 | 152 | ||
| CR541937.1 | - | 1 | 297 | 152 | ||
| CR541962.1 | - | 1 | 297 | 152 | ||
| U27467.1 | - | 1 | 331 | 152 | ||
| U29680.1 | - | 1 | 375 | 152 | ||
| Y09397.1 | - | 1 | 399 | 152 | ||
More Information
Set Membership: |
|

BCL2A1
Genomic Map