Assay Details
| Gene Symbol: SPN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030288.2 | NP_001025459.1 | 2 | 758 | 85 | |
| NM_003123.4 | NP_003114.1 | 2 | 725 | 85 | ||
| GenBank mRNA | AK292626.1 | - | 2 | 697 | 85 | |
| AK313750.1 | - | 2 | 697 | 85 | ||
| BC012350.1 | - | 2 | 645 | 4564347 | 85 | |
| J04536.1 | - | 2 | 685 | 85 | ||
| X60502.1 | - | 2 | 729 | 85 | ||
More Information
Set Membership: |
|

SPN
Genomic Map