Assay Details
| Gene Symbol: EPPIN-WFDC6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198986.1 | NP_001185915.1 | 5 | 1249 | 115 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

EPPIN-WFDC6| Gene Symbol: EPPIN-WFDC6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198986.1 | NP_001185915.1 | 5 | 1249 | 115 | |
Set Membership: |
|