Assay Details
| Gene Symbol: WDR6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001320546.1 | NP_001307475.1 | 2 | 2857 | 96 | |
| NM_001320547.1 | NP_001307476.1 | 2 | 2228 | 96 | ||
| NM_018031.4 | NP_060501.3 | 2 | 2643 | 96 | ||
| GenBank mRNA | AF099100.1 | - | 2 | 2312 | 96 | |
| AK022719.1 | - | 1 | 548 | 96 | ||
| AK225653.1 | - | 2 | 2306 | 96 | ||
| AK295145.1 | - | 2 | 2643 | 96 | ||
| AK297090.1 | - | 2 | 653 | 96 | ||
| AK304117.1 | - | 2 | 2228 | 96 | ||
| AK309913.1 | - | 2 | 2523 | 96 | ||
| AL133589.1 | - | 1 | 423 | 96 | ||
| AL833910.1 | - | 1 | 1489 | 96 | ||
| BC078176.1 | - | 1 | 362 | 96 | ||
| BC101707.1 | - | 2 | 2302 | 8069213 | 96 | |
| BC113467.1 | - | 2 | 2302 | 8322519 | 96 | |
| BC143486.1 | - | 2 | 2302 | 96 | ||
More Information
Set Membership: |
|

WDR6
Genomic Map