Assay Details
| Gene Symbol: THG1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001317825.1 | NP_001304754.1 | 3 | 454 | 65 | |
| NM_001317826.1 | NP_001304755.1 | 4 | 426 | 65 | ||
| NM_017872.4 | NP_060342.2 | 3 | 458 | 65 | ||
| XM_017009603.1 | XP_016865092.1 | 3 | 393 | 65 | ||
| GenBank mRNA | AK000553.1 | - | 3 | 434 | 65 | |
| AK021663.1 | - | 3 | 409 | 65 | ||
| AK023355.1 | - | 2 | 72 | 65 | ||
| AK223119.1 | - | 3 | 427 | 65 | ||
| AL136669.1 | - | 3 | 426 | 65 | ||
| AY463216.1 | - | 3 | 532 | 65 | ||
| BC001523.2 | - | 3 | 404 | 3010614 | 65 | |
| BC001852.2 | - | 3 | 404 | 3010614 | 65 | |
| BC023521.2 | - | 3 | 424 | 4126840 | 65 | |
| CR533503.1 | - | 3 | 406 | 65 | ||
| DA874954.1 | - | 3 | 421 | 65 | ||
More Information
Set Membership: |
|

THG1L
Genomic Map