Assay Details
| Gene Symbol: SERPINB6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017010941.1 | XP_016866430.1 | 1 | 332 | 61 | |
| GenBank mRNA | BX538343.1 | - | 1 | 361 | 61 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BX538343.1, XM_017010941.1 transcripts.
Set Membership: |
|

SERPINB6
Genomic Map