Assay Details
| Gene Symbol: LOC105373132 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011544354.2 | XP_011542656.1 | 1 | 1676 | 77 | |
| Gene Symbol: DUSP5P1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK055963.1 | - | 1 | 346 | 77 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK055963.1, XM_011544354.2 transcripts.
Set Membership: |
|

LOC105373132
Genomic Map