Assay Details
| Gene Symbol: FMR1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | EU048200.1 | - | 2 | 578 | 54 | |
| EU048201.1 | - | 2 | 578 | 54 | ||
| EU048202.1 | - | 2 | 578 | 54 | ||
| EU048203.1 | - | 2 | 578 | 54 | ||
| EU048204.1 | - | 3 | 494 | 54 | ||
More Information
Set Membership: |
|

FMR1-AS1
Genomic Map