Assay Details
| Gene Symbol: TPT1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK124332.1 | - | 5 | 717 | 97 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK124332.1 transcripts.
Set Membership: |
|

TPT1-AS1
Genomic Map