Assay Details
Gene Symbol: ZNF780B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005258590.4 | XP_005258647.1 | 1 - 2 | 87 | 63 | |
XM_005258593.2 | XP_005258650.1 | 1 - 2 | 74 | 63 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005258590.4, XM_005258593.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
