Assay Details
| Gene Symbol: TAF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031923.3 | NP_114129.1 | 2 - 3 | 617 | 74 | |
| GenBank mRNA | AJ292190.1 | - | 2 - 3 | 411 | 74 | |
| AK307657.1 | - | 2 - 3 | 595 | 74 | ||
| AL117661.1 | - | 2 - 3 | 345 | 74 | ||
| BC017320.1 | - | 2 - 3 | 603 | 74 | ||
| BC028077.1 | - | 2 - 3 | 574 | 74 | ||
| BC045106.1 | - | 2 - 3 | 603 | 74 | ||
| BC062352.1 | - | 2 - 3 | 584 | 74 | ||
| BC073884.1 | - | 2 - 3 | 608 | 74 | ||
| BC105593.1 | - | 2 - 3 | 621 | 74 | ||
More Information
Set Membership: |
|

TAF3
Genomic Map