Assay Details
Gene Symbol: PRORY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001282471.1 | NP_001269400.1 | 3 - 4 | 548 | 86 | |
GenBank mRNA | AK026367.1 | - | 4 - 5 | 747 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
