Assay Details
| Gene Symbol: TRIB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001282985.1 | NP_001269914.1 | 2 - 3 | 414 | 115 | |
| NM_025195.3 | NP_079471.1 | 2 - 3 | 1240 | 115 | ||
| GenBank mRNA | AF205437.1 | - | 2 - 3 | 931 | 115 | |
| AF250310.1 | - | 2 - 3 | 939 | 115 | ||
| AJ000480.1 | - | 1 - 2 | 214 | 115 | ||
| AK297539.1 | - | 2 - 3 | 355 | 115 | ||
| AL832388.1 | - | 2 - 3 | 872 | 115 | ||
| BC063292.1 | - | 2 - 3 | 1240 | 30343492 | 115 | |
| DA686993.1 | - | 2 - 3 | 414 | 115 | ||
More Information
Set Membership: |
|

TRIB1
Genomic Map