Assay Details
| Gene Symbol: MED30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001282986.1 | NP_001269915.1 | 2 - 3 | 499 | 105 | |
| GenBank mRNA | FM179284.1 | - | 2 - 3 | 379 | 105 | |
More Information
Set Membership: |
|

MED30
Genomic Map