Assay Details
| Gene Symbol: NME4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286433.1 | NP_001273362.1 | 2 - 3 | 261 | 88 | |
| GenBank mRNA | HY182358.1 | - | 2 - 3 | 188 | 88 | |
More Information
Set Membership: |
|

NME4
Genomic Map