Assay Details
| Gene Symbol: GLIPR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001287010.1 | NP_001273939.1 | 1 - 2 | 251 | 100 | |
| NM_001287011.1 | NP_001273940.1 | 1 - 2 | 251 | 100 | ||
| NM_001287012.1 | NP_001273941.1 | 1 - 2 | 251 | 100 | ||
| NM_022343.3 | NP_071738.1 | 1 - 2 | 251 | 100 | ||
| NR_104639.1 | 1 - 2 | 251 | 100 | |||
| NR_104640.1 | 1 - 2 | 251 | 100 | |||
| GenBank mRNA | AK074488.1 | - | 1 - 2 | 73 | 100 | |
| BF515272.1 | - | 1 - 2 | 251 | 100 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK074488.1, NM_001287010.1, NM_001287011.1, NM_001287012.1, NM_022343.3 transcripts.
Set Membership: |
|

GLIPR2
Genomic Map