Assay Details
| Gene Symbol: GLIPR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001287012.1 | NP_001273941.1 | 2 - 3 | 364 | 86 | |
| GenBank mRNA | DA432700.1 | - | 2 - 3 | 165 | 86 | |
More Information
Set Membership: |
|

GLIPR2
Genomic Map