Assay Details
| Gene Symbol: PHPT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001287342.1 | NP_001274271.1 | 2 - 3 | 904 | 92 | |
| NM_014172.5 | NP_054891.2 | 2 - 3 | 898 | 92 | ||
| NR_109808.1 | 2 - 3 | 881 | 92 | |||
| GenBank mRNA | AA460844.1 | - | 1 - 2 | 309 | 93 | |
| AF161490.1 | - | 2 - 3 | 303 | 92 | ||
| AF164795.1 | - | 2 - 3 | 291 | 92 | ||
| AF285119.1 | - | 2 - 3 | 616 | 92 | ||
| AF393504.1 | - | 2 - 3 | 283 | 92 | ||
| AL136644.1 | - | 2 - 3 | 308 | 92 | ||
| BC024648.1 | - | 2 - 3 | 304 | 4934238 | 92 | |
| BP375594.1 | - | 2 - 3 | 302 | 92 | ||
| CD693554.1 | - | 2 - 3 | 343 | 92 | ||
| CN334799.1 | - | 2 - 3 | 281 | 92 | ||
| CR533481.1 | - | 2 - 3 | 283 | 92 | ||
| EU794646.1 | - | 2 - 3 | 615 | 92 | ||
| F29783.1 | - | 2 - 3 | 141 | 92 | ||
More Information
Set Membership: |
|

PHPT1
Genomic Map