Assay Details
| Gene Symbol: SLC50A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001287589.1 | NP_001274518.1 | 2 - 3 | 256 | 76 | |
| NM_001287592.1 | NP_001274521.1 | 2 - 3 | 331 | 76 | ||
| GenBank mRNA | BX093804.1 | - | 2 - 3 | 248 | 76 | |
| X84712.1 | - | 2 - 3 | 127 | 76 | ||
More Information
Set Membership: |
|

SLC50A1
Genomic Map