Assay Details
| Gene Symbol: ZKSCAN7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001288592.1 | NP_001275521.1 | 1 - 2 | 303 | 97 | |
| GenBank mRNA | BC148260.1 | - | 1 - 2 | 34 | 97 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001288592.1 transcripts.
Set Membership: |
|

ZKSCAN7
Genomic Map