Assay Details
| Gene Symbol: ZKSCAN7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001288592.1 | NP_001275521.1 | 4 - 5 | 697 | 87 | |
| GenBank mRNA | AK022800.1 | - | 4 - 5 | 1092 | 87 | |
| BC148260.1 | - | 4 - 5 | 428 | 87 | ||
More Information
Set Membership: |
|

ZKSCAN7
Genomic Map